Assay Details
| Gene Symbol: Tjp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282095.1 | NP_001269024.1 | 17 - 18 | 2322 | 72 | |
| NM_001282096.1 | NP_001269025.1 | 17 - 18 | 2385 | 72 | ||
| NM_013769.3 | NP_038797.2 | 17 - 18 | 2424 | 72 | ||
| GenBank mRNA | AF157006.1 | - | 17 - 18 | 2395 | 72 | |
| AK008855.1 | - | 5 - 6 | 777 | 72 | ||
| AK029083.1 | - | 17 - 18 | 2371 | 72 | ||
| AK134100.1 | - | 17 - 18 | 2425 | 72 | ||
| BC012518.1 | - | 17 - 18 | 2389 | 3600848 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tjp3
Genomic Map