Assay Details
| Gene Symbol: Matk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285853.1 | NP_001272782.1 | 11 - 12 | 1404 | 79 | |
| NM_001285854.1 | NP_001272783.1 | 11 - 12 | 1401 | 79 | ||
| NM_001285855.1 | NP_001272784.1 | 11 - 12 | 1469 | 79 | ||
| NM_010768.2 | NP_034898.1 | 12 - 13 | 1537 | 79 | ||
| GenBank mRNA | AK013606.1 | - | 11 - 12 | 1218 | 79 | |
| AK158883.1 | - | 8 - 9 | 1339 | 79 | ||
| BC060086.1 | - | 7 - 8 | 2536 | 79 | ||
| BC111864.1 | - | 9 - 10 | 1386 | 40046807 | 79 | |
| D45243.1 | - | 11 - 12 | 1469 | 79 | ||
| L27738.1 | - | 12 - 13 | 1291 | 79 | ||
| S77473.1 | - | 11 - 12 | 1216 | 79 | ||
| U05210.1 | - | 11 - 12 | 1190 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Matk
Genomic Map