Assay Details
| Gene Symbol: Ncln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134009.3 | NP_598770.1 | 4 - 5 | 718 | 87 | |
| GenBank mRNA | AK019409.1 | - | 4 - 5 | 588 | 87 | |
| AK048954.1 | - | 4 - 5 | 712 | 87 | ||
| AK128887.1 | - | 4 - 5 | 719 | 87 | ||
| BC019501.1 | - | 4 - 5 | 718 | 4193809 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncln
Genomic Map