Assay Details
| Gene Symbol: Sppl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175195.3 | NP_780404.2 | 13 - 14 | 1401 | 80 | |
| GenBank mRNA | AK148109.1 | - | 13 - 14 | 1368 | 80 | |
| AK165324.1 | - | 12 - 13 | 1426 | 80 | ||
| AK166364.1 | - | 13 - 14 | 1352 | 80 | ||
| AK170375.1 | - | 13 - 14 | 1367 | 80 | ||
| AK201100.1 | - | 2 - 3 | 123 | 80 | ||
| AK207842.1 | - | 3 - 4 | 232 | 80 | ||
| AK208656.1 | - | 2 - 3 | 114 | 80 | ||
| BC034206.1 | - | 10 - 11 | 956 | 80 | ||
| BC052094.1 | - | 11 - 12 | 1075 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sppl2b
Genomic Map