Assay Details
| Gene Symbol: Sppl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175195.3 | NP_780404.2 | 3 - 4 | 417 | 79 | |
| GenBank mRNA | AK046804.1 | - | 3 - 4 | 436 | 79 | |
| AK148109.1 | - | 3 - 4 | 384 | 79 | ||
| AK157906.1 | - | 3 - 4 | 362 | 79 | ||
| AK165324.1 | - | 3 - 4 | 539 | 79 | ||
| AK166364.1 | - | 3 - 4 | 368 | 79 | ||
| AK170375.1 | - | 3 - 4 | 380 | 79 | ||
| AK197828.1 | - | 1 - 2 | 87 | 79 | ||
| BC052094.1 | - | 1 - 2 | 91 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046804.1 transcripts.
Set Membership: |
|

Sppl2b
Genomic Map