Assay Details
| Gene Symbol: Lrrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146048.1 | NP_001139520.1 | 10 - 11 | 1221 | 88 | |
| NM_172528.3 | NP_766116.3 | 10 - 11 | 869 | 88 | ||
| GenBank mRNA | AK082721.1 | - | 10 - 11 | 870 | 88 | |
| AK147075.1 | - | 10 - 11 | 1221 | 88 | ||
| AK220166.1 | - | 10 - 11 | 1239 | 88 | ||
| BC022613.1 | - | 2 - 3 | 163 | 88 | ||
| BC046591.1 | - | 10 - 11 | 1208 | 88 | ||
| BC087542.1 | - | 10 - 11 | 1068 | 4934980 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc1
Genomic Map