| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900022348.mm.1 |
NM_001146048.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022348.mm.1 |
NM_001146048.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022348.mm.1 |
TC0900002702.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900022350.mm.1 |
NM_001146048.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022350.mm.1 |
NM_001146048.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022350.mm.1 |
TC0900002702.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900022341.mm.1 |
NM_001146048.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022341.mm.1 |
NM_001146048.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022341.mm.1 |
NM_172528.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022341.mm.1 |
NM_172528.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022341.mm.1 |
TC0900002702.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900022349.mm.1 |
NM_001146048.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022349.mm.1 |
NM_001146048.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022349.mm.1 |
TC0900002702.mm.1 |
MTA TC |