Assay Details
| Gene Symbol: Fbxo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081490.2 | NP_001074959.1 | 4 - 5 | 365 | 71 | |
| NM_023605.2 | NP_076094.2 | 4 - 5 | 667 | 71 | ||
| GenBank mRNA | AB041585.1 | - | 4 - 5 | 636 | 71 | |
| AK018482.1 | - | 4 - 5 | 618 | 71 | ||
| AK077607.1 | - | 4 - 5 | 328 | 71 | ||
| AK182638.1 | - | 3 - 4 | 257 | 71 | ||
| BC020074.1 | - | 4 - 5 | 638 | 3710611 | 71 | |
| DV657928.1 | - | 4 - 5 | 353 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo9
Genomic Map