Assay Details
| Gene Symbol: Fbxo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081490.2 | NP_001074959.1 | 6 - 7 | 596 | 81 | |
| NM_023605.2 | NP_076094.2 | 6 - 7 | 898 | 81 | ||
| GenBank mRNA | AB041585.1 | - | 6 - 7 | 867 | 81 | |
| AK077607.1 | - | 6 - 7 | 559 | 81 | ||
| BC020074.1 | - | 6 - 7 | 869 | 3710611 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo9
Genomic Map