Assay Details
| Gene Symbol: Fbxo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081490.2 | NP_001074959.1 | 8 - 9 | 824 | 82 | |
| NM_023605.2 | NP_076094.2 | 8 - 9 | 1126 | 82 | ||
| GenBank mRNA | AB041585.1 | - | 8 - 9 | 1096 | 82 | |
| AK077607.1 | - | 8 - 9 | 787 | 82 | ||
| BC020074.1 | - | 8 - 9 | 1097 | 3710611 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo9
Genomic Map