Assay Details
| Gene Symbol: Fbxo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081490.2 | NP_001074959.1 | 9 - 10 | 921 | 62 | |
| NM_023605.2 | NP_076094.2 | 9 - 10 | 1223 | 62 | ||
| GenBank mRNA | AB041585.1 | - | 9 - 10 | 1193 | 62 | |
| AK077607.1 | - | 9 - 10 | 884 | 62 | ||
| BC020074.1 | - | 9 - 10 | 1194 | 3710611 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo9
Genomic Map