Assay Details
| Gene Symbol: Cdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007659.3 | NP_031685.2 | 6 - 7 | 757 | 73 | |
| GenBank mRNA | AK030231.1 | - | 6 - 7 | 758 | 73 | |
| AK051859.1 | - | 6 - 7 | 784 | 73 | ||
| AK135516.1 | - | 6 - 7 | 761 | 73 | ||
| AK168054.1 | - | 6 - 7 | 758 | 73 | ||
| BC005614.1 | - | 5 - 6 | 650 | 73 | ||
| BC024396.1 | - | 6 - 7 | 735 | 4977366 | 73 | |
| M38724.1 | - | 5 - 6 | 678 | 73 | ||
| X16461.1 | - | 5 - 6 | 678 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk1
Genomic Map