Assay Details
| Gene Symbol: Yif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110201.1 | NP_001103671.1 | 2 - 3 | 351 | 81 | |
| NM_029887.3 | NP_084163.2 | 2 - 3 | 360 | 81 | ||
| GenBank mRNA | AK020444.1 | - | 2 - 3 | 331 | 81 | |
| AK135529.1 | - | 329 | 81 | |||
| BC109331.2 | - | 2 - 3 | 40051521 | 81 | ||
| BU921944.1 | - | 2 - 3 | 282 | 81 | ||
| BU953116.1 | - | 2 - 3 | 348 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Yif1b
Genomic Map