Assay Details
| Gene Symbol: Sirt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122765.1 | NP_001116237.1 | 11 - 12 | 875 | 116 | |
| NM_001122766.1 | NP_001116238.1 | 9 - 10 | 712 | 116 | ||
| NM_022432.4 | NP_071877.3 | 12 - 13 | 922 | 116 | ||
| GenBank mRNA | AB221570.1 | - | 12 - 13 | 827 | 116 | |
| AF299337.1 | - | 12 - 13 | 907 | 116 | ||
| AK014042.1 | - | 11 - 12 | 846 | 116 | ||
| AK144431.1 | - | 11 - 12 | 819 | 116 | ||
| AK146412.1 | - | 11 - 12 | 852 | 116 | ||
| BC021439.1 | - | 12 - 13 | 926 | 5039278 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sirt2
Genomic Map