Assay Details
| Gene Symbol: Kcnn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163510.1 | NP_001156982.1 | 4 - 5 | 989 | 153 | |
| NM_008433.4 | NP_032459.3 | 4 - 5 | 989 | 153 | ||
| GenBank mRNA | AB128930.1 | - | 2 - 3 | 752 | 153 | |
| AF042487.1 | - | 4 - 5 | 855 | 153 | ||
| AF072884.1 | - | 4 - 5 | 815 | 153 | ||
| AK010943.1 | - | 4 - 5 | 990 | 153 | ||
| AK085752.1 | - | 4 - 5 | 806 | 153 | ||
| AK155910.1 | - | 4 - 5 | 954 | 153 | ||
| BC010274.1 | - | 4 - 5 | 938 | 3589411 | 153 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB128930.1 transcripts.
Set Membership: |
|

Kcnn4
Genomic Map