Assay Details
| Gene Symbol: Timm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025616.3 | NP_079892.1 | 6 - 7 | 511 | 83 | |
| GenBank mRNA | AK008340.1 | - | 6 - 7 | 512 | 83 | |
| AK008382.1 | - | 6 - 7 | 509 | 83 | ||
| AK010832.1 | - | 6 - 7 | 477 | 83 | ||
| AK012977.1 | - | 6 - 7 | 508 | 83 | ||
| AK088074.1 | - | 6 - 7 | 504 | 83 | ||
| AK167473.1 | - | 6 - 7 | 514 | 83 | ||
| AY551342.1 | - | 6 - 7 | 497 | 83 | ||
| BC010303.1 | - | 6 - 7 | 486 | 3597676 | 83 | |
| BC037056.1 | - | 6 - 7 | 513 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timm50
Genomic Map