Assay Details
| Gene Symbol: Timm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025616.3 | NP_079892.1 | 7 - 8 | 614 | 83 | |
| GenBank mRNA | AK008340.1 | - | 7 - 8 | 615 | 83 | |
| AK008382.1 | - | 7 - 8 | 612 | 83 | ||
| AK010832.1 | - | 7 - 8 | 580 | 83 | ||
| AK088074.1 | - | 7 - 8 | 607 | 83 | ||
| AK167473.1 | - | 7 - 8 | 617 | 83 | ||
| AK186280.1 | - | 2 - 3 | 136 | 83 | ||
| AY551342.1 | - | 7 - 8 | 600 | 83 | ||
| BC010303.1 | - | 7 - 8 | 589 | 3597676 | 83 | |
| BC025844.1 | - | 1 - 2 | 95 | 83 | ||
| BC037056.1 | - | 7 - 8 | 616 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timm50
Genomic Map