Assay Details
| Gene Symbol: Timm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025616.3 | NP_079892.1 | 8 - 9 | 707 | 62 | |
| GenBank mRNA | AK008340.1 | - | 8 - 9 | 708 | 62 | |
| AK008382.1 | - | 8 - 9 | 705 | 62 | ||
| AK010832.1 | - | 8 - 9 | 673 | 62 | ||
| AK167473.1 | - | 8 - 9 | 710 | 62 | ||
| AK187841.1 | - | 3 - 4 | 339 | 62 | ||
| AY551342.1 | - | 8 - 9 | 693 | 62 | ||
| BC010303.1 | - | 8 - 9 | 682 | 3597676 | 62 | |
| BC025844.1 | - | 2 - 3 | 188 | 62 | ||
| BC037056.1 | - | 8 - 9 | 709 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timm50
Genomic Map