Assay Details
| Gene Symbol: Atp1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290469.1 | NP_001277398.1 | 3 - 4 | 327 | 64 | |
| GenBank mRNA | AK053751.1 | - | 3 - 4 | 299 | 64 | |
| BC020177.1 | - | 3 - 4 | 285 | 4506376 | 64 | |
| BC034645.1 | - | 3 - 4 | 283 | 5357731 | 64 | |
| BC037206.1 | - | 3 - 4 | 282 | 5362423 | 64 | |
| BC042894.1 | - | 3 - 4 | 327 | 5364812 | 64 | |
More Information
Set Membership: |
|

Atp1a3
Genomic Map