Assay Details
| Gene Symbol: Atp1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290469.1 | NP_001277398.1 | 9 - 10 | 1368 | 61 | |
| GenBank mRNA | AK053751.1 | - | 9 - 10 | 1340 | 61 | |
| BC020177.1 | - | 9 - 10 | 1326 | 4506376 | 61 | |
| BC027000.1 | - | 2 - 3 | 388 | 61 | ||
| BC034645.1 | - | 9 - 10 | 1324 | 5357731 | 61 | |
| BC037206.1 | - | 9 - 10 | 1323 | 5362423 | 61 | |
| BC042894.1 | - | 9 - 10 | 1368 | 5364812 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a3
Genomic Map