Assay Details
| Gene Symbol: Adamtsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285435.1 | NP_001272364.1 | 10 - 11 | 1107 | 108 | |
| NR_104339.1 | 9 - 10 | 1625 | 108 | |||
| NR_104340.1 | 9 - 10 | 1622 | 108 | |||
| GenBank mRNA | AK028819.1 | - | 10 - 11 | 1579 | 108 | |
| AK029081.1 | - | 9 - 10 | 1563 | 108 | ||
| AK154947.1 | - | 6 - 7 | 1212 | 108 | ||
| BC034843.1 | - | 8 - 9 | 1266 | 108 | ||
| BC058577.1 | - | 5 - 6 | 2198 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adamtsl5
Genomic Map