Assay Details
| Gene Symbol: Adarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130895.3 | NP_570965.2 | 7 - 8 | 1789 | 77 | |
| NR_021486.1 | 7 - 8 | 1836 | 77 | |||
| GenBank mRNA | AF291049.1 | - | 6 - 7 | 1624 | 77 | |
| AF403106.2 | - | 7 - 8 | 1741 | 77 | ||
| AF403108.2 | - | 7 - 8 | 1788 | 77 | ||
| AF525421.1 | - | 8 - 9 | 1814 | 77 | ||
| AY162454.1 | - | 8 - 9 | 1814 | 77 | ||
More Information
Set Membership: |
|

Adarb1
Genomic Map