Assay Details
| Gene Symbol: Adarb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004429.1 | 3 - 4 | 428 | 99 | ||
| NR_021486.1 | 3 - 4 | 428 | 99 | |||
| GenBank mRNA | AF403108.2 | - | 3 - 4 | 380 | 99 | |
| AF403109.2 | - | 3 - 4 | 380 | 99 | ||
| BB622419.1 | - | 3 - 4 | 428 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein O-fucosyltransferase 2 | Pofut2 |
2310011G23Rik, AI256847, BC003494, C21orf80, FUT13 |
-52.1kb | Anti-sense | Chr. 10 - 77259300 - 77269586 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 17769 |
Gm17769 | 92.4kb | Anti-sense | Chr. 10 - 77418376 - 77420939 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF403108.2, AF403109.2 transcripts.
Set Membership: |
|

Adarb1
Genomic Map