Assay Details
| Gene Symbol: Pfkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008826.4 | NP_032852.2 | 14 - 15 | 1528 | 86 | |
| GenBank mRNA | AK030511.1 | - | 14 - 15 | 1483 | 86 | |
| AK030892.1 | - | 7 - 8 | 627 | 86 | ||
| AK036318.1 | - | 14 - 15 | 1461 | 86 | ||
| AK081313.1 | - | 14 - 15 | 1479 | 86 | ||
| AK159328.1 | - | 14 - 15 | 1473 | 86 | ||
| AK168660.1 | - | 14 - 15 | 1484 | 86 | ||
| BC020097.1 | - | 14 - 15 | 1482 | 4014395 | 86 | |
| BC029336.1 | - | 14 - 15 | 1471 | 86 | ||
| J03928.1 | - | 14 - 15 | 1457 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkl
Genomic Map