Assay Details
| Gene Symbol: Aire | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271549.1 | NP_001258478.1 | 7 - 8 | 974 | 61 | |
| NM_001271551.1 | NP_001258480.1 | 7 - 8 | 962 | 61 | ||
| NM_001271553.1 | NP_001258482.1 | 7 - 8 | 974 | 61 | ||
| NM_001271555.1 | NP_001258484.1 | 7 - 8 | 962 | 61 | ||
| NM_001271557.1 | NP_001258486.1 | 7 - 8 | 974 | 61 | ||
| NM_001271559.1 | NP_001258488.1 | 7 - 8 | 962 | 61 | ||
| GenBank mRNA | AF128115.1 | - | 7 - 8 | 944 | 61 | |
| AF128117.1 | - | 7 - 8 | 932 | 61 | ||
| AF128119.1 | - | 7 - 8 | 944 | 61 | ||
| AF128121.1 | - | 7 - 8 | 932 | 61 | ||
| AF128123.1 | - | 7 - 8 | 944 | 61 | ||
| AF128125.1 | - | 7 - 8 | 932 | 61 | ||
| AK153690.1 | - | 8 - 9 | 1028 | 61 | ||
| BC103518.1 | - | 7 - 8 | 936 | 40039058 | 61 | |
More Information
Set Membership: |
|

Aire
Genomic Map