Assay Details
| Gene Symbol: Pcbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021568.2 | NP_067543.2 | 14 - 15 | 1339 | 92 | |
| GenBank mRNA | AF176327.1 | - | 14 - 15 | 1180 | 92 | |
| AK034811.1 | - | 17 - 18 | 1715 | 92 | ||
| AK079564.1 | - | 12 - 13 | 1381 | 92 | ||
| BC042440.1 | - | 10 - 11 | 904 | 4511401 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcbp3
Genomic Map