Assay Details
| Gene Symbol: Derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024440.2 | NP_077760.1 | 1 - 2 | 122 | 90 | |
| GenBank mRNA | AB047555.1 | - | 1 - 2 | 122 | 90 | |
| AK007348.1 | - | 1 - 2 | 107 | 90 | ||
| AK007948.1 | - | 1 - 2 | 108 | 90 | ||
| AK172429.1 | - | 1 - 2 | 108 | 90 | ||
| BC004729.1 | - | 1 - 2 | 105 | 90 | ||
| BC019815.1 | - | 1 - 2 | 101 | 90 | ||
| BC115466.1 | - | 1 - 2 | 117 | 40100387 | 90 | |
| BC115467.1 | - | 1 - 2 | 117 | 40100388 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Derl3
Genomic Map