Assay Details
| Gene Symbol: Ggt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011820.4 | NP_035950.2 | 1 - 2 | 502 | 107 | |
| GenBank mRNA | AF077765.1 | - | 1 - 2 | 487 | 107 | |
| AK049084.1 | - | 1 - 2 | 503 | 108 | ||
| AK052183.1 | - | 1 - 2 | 503 | 107 | ||
| AK085788.1 | - | 1 - 2 | 502 | 109 | ||
| AK140308.1 | - | 1 - 2 | 543 | 107 | ||
| AK170326.1 | - | 1 - 2 | 366 | 107 | ||
| BC051971.1 | - | 1 - 2 | 282 | 107 | ||
| BC113775.1 | - | 1 - 2 | 226 | 40059928 | 107 | |
More Information
Set Membership: |
|

Ggt5
Genomic Map