Assay Details
| Gene Symbol: Ggt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011820.4 | NP_035950.2 | 10 - 11 | 1785 | 86 | |
| GenBank mRNA | AK029138.1 | - | 3 - 4 | 321 | 86 | |
| AK049084.1 | - | 10 - 11 | 1789 | 86 | ||
| AK052183.1 | - | 10 - 11 | 1786 | 86 | ||
| AK140308.1 | - | 9 - 10 | 2306 | 86 | ||
| AK170326.1 | - | 10 - 11 | 1649 | 86 | ||
| BC051971.1 | - | 9 - 10 | 1480 | 86 | ||
| BC089585.1 | - | 6 - 7 | 1204 | 86 | ||
| BC113775.1 | - | 10 - 11 | 1509 | 40059928 | 86 | |
More Information
Set Membership: |
|

Ggt5
Genomic Map