Assay Details
| Gene Symbol: Upb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133995.4 | NP_598756.1 | 5 - 6 | 766 | 67 | |
| GenBank mRNA | AK149373.1 | - | 5 - 6 | 663 | 67 | |
| AK149480.1 | - | 5 - 6 | 663 | 67 | ||
| BC021388.1 | - | 5 - 6 | 651 | 5055165 | 67 | |
| BC024447.1 | - | 3 - 4 | 317 | 67 | ||
More Information
Set Membership: |
|

Upb1
Genomic Map