Assay Details
| Gene Symbol: Upb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133995.4 | NP_598756.1 | 7 - 8 | 1012 | 89 | |
| GenBank mRNA | AK149373.1 | - | 7 - 8 | 909 | 89 | |
| AK149480.1 | - | 7 - 8 | 909 | 89 | ||
| BC021388.1 | - | 7 - 8 | 897 | 5055165 | 89 | |
| BC024447.1 | - | 5 - 6 | 563 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Upb1
Genomic Map