Assay Details
| Gene Symbol: Tbata | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017419.2 | NP_001017419.1 | 9 - 10 | 1185 | 65 | |
| NM_001017433.2 | NP_001017433.1 | 9 - 10 | 1287 | 65 | ||
| GenBank mRNA | AF521591.1 | - | 8 - 9 | 1116 | 65 | |
| AF521592.1 | - | 8 - 9 | 1218 | 65 | ||
| AK006213.1 | - | 8 - 9 | 1137 | 65 | ||
| AW490515.1 | - | 1 - 2 | 65 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tbata
Genomic Map