Assay Details
Gene Symbol: Izumo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027829.3 | NP_082105.3 | 1 - 2 | 269 | 67 | |
GenBank mRNA | BC057953.1 | - | 1 - 2 | 195 | 67 | |
BF682661.1 | - | 1 - 2 | 204 | 67 | ||
CF105977.1 | - | 1 - 2 | 269 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC057953.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
