Assay Details
| Gene Symbol: Plk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183152.3 | NP_898975.2 | 11 - 12 | 1312 | 89 | |
| GenBank mRNA | AK031968.1 | - | 11 - 12 | 1313 | 89 | |
| BC086684.1 | - | 6 - 7 | 609 | 30679265 | 89 | |
| BC099679.1 | - | 11 - 12 | 1281 | 6489647 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plk5
Genomic Map