Assay Details
| Gene Symbol: Plk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183152.3 | NP_898975.2 | 2 - 3 | 318 | 91 | |
| GenBank mRNA | BB622163.1 | - | 2 - 3 | 319 | 91 | |
| BC099679.1 | - | 2 - 3 | 299 | 6489647 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plk5
Genomic Map