Assay Details
| Gene Symbol: Plk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183152.3 | NP_898975.2 | 7 - 8 | 958 | 76 | |
| GenBank mRNA | AK031968.1 | - | 7 - 8 | 959 | 76 | |
| BC086684.1 | - | 2 - 3 | 255 | 30679265 | 76 | |
| BC099679.1 | - | 7 - 8 | 927 | 6489647 | 76 | |
| BE654673.1 | - | 3 - 4 | 341 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plk5
Genomic Map