Assay Details
| Gene Symbol: Plk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183152.3 | NP_898975.2 | 9 - 10 | 1159 | 61 | |
| GenBank mRNA | AK031968.1 | - | 9 - 10 | 1160 | 61 | |
| BC086684.1 | - | 4 - 5 | 456 | 30679265 | 61 | |
| BC099679.1 | - | 9 - 10 | 1128 | 6489647 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plk5
Genomic Map