Assay Details
| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029781.3 | NP_084057.1 | 2 - 3 | 320 | 74 | |
| GenBank mRNA | AB232632.1 | - | 1 - 2 | 75 | 74 | |
| AK018269.1 | - | 2 - 3 | 339 | 74 | ||
| AK038445.1 | - | 2 - 3 | 321 | 74 | ||
| AK043946.1 | - | 2 - 3 | 344 | 74 | ||
| AK046113.1 | - | 2 - 3 | 129 | 74 | ||
| AK142025.1 | - | 1 - 2 | 692 | 74 | ||
| AK142415.1 | - | 1 - 2 | 1302 | 74 | ||
| BC119094.1 | - | 2 - 3 | 308 | 8733843 | 74 | |
| BC119098.1 | - | 2 - 3 | 295 | 8733847 | 74 | |
More Information
Set Membership: |
|

Rab36
Genomic Map