Assay Details
| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029781.3 | NP_084057.1 | 4 - 5 | 472 | 72 | |
| GenBank mRNA | AB232632.1 | - | 3 - 4 | 227 | 72 | |
| AK018269.1 | - | 4 - 5 | 491 | 72 | ||
| AK038445.1 | - | 4 - 5 | 473 | 72 | ||
| AK046113.1 | - | 4 - 5 | 281 | 72 | ||
| AK142025.1 | - | 3 - 4 | 844 | 72 | ||
| AK142415.1 | - | 3 - 4 | 1454 | 72 | ||
| AK163455.1 | - | 4 - 5 | 480 | 72 | ||
| BC119094.1 | - | 4 - 5 | 460 | 8733843 | 72 | |
| BC119098.1 | - | 4 - 5 | 447 | 8733847 | 72 | |
More Information
Set Membership: |
|

Rab36
Genomic Map