Assay Details
| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029781.3 | NP_084057.1 | 5 - 6 | 575 | 81 | |
| GenBank mRNA | AB232632.1 | - | 4 - 5 | 330 | 81 | |
| AK018269.1 | - | 5 - 6 | 594 | 81 | ||
| AK038445.1 | - | 5 - 6 | 576 | 81 | ||
| AK043946.1 | - | 5 - 6 | 599 | 81 | ||
| AK046113.1 | - | 5 - 6 | 384 | 81 | ||
| AK142025.1 | - | 4 - 5 | 947 | 81 | ||
| AK163455.1 | - | 5 - 6 | 583 | 81 | ||
| BC119094.1 | - | 5 - 6 | 563 | 8733843 | 81 | |
| BC119098.1 | - | 5 - 6 | 550 | 8733847 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab36
Genomic Map