Assay Details
| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029781.3 | NP_084057.1 | 7 - 8 | 695 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab36| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029781.3 | NP_084057.1 | 7 - 8 | 695 | 70 | |
Set Membership: |
|