Assay Details
| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029781.3 | NP_084057.1 | 9 - 10 | 860 | 75 | |
| GenBank mRNA | AB232632.1 | - | 8 - 9 | 615 | 75 | |
| AK038445.1 | - | 9 - 10 | 861 | 75 | ||
| AK046113.1 | - | 9 - 10 | 669 | 75 | ||
| AK142025.1 | - | 8 - 9 | 1232 | 75 | ||
| BC119094.1 | - | 9 - 10 | 848 | 8733843 | 75 | |
| BC119098.1 | - | 9 - 10 | 835 | 8733847 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab36
Genomic Map