Assay Details
| Gene Symbol: Aifm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039194.3 | NP_001034283.1 | 1 - 2 | 92 | 59 | |
| NM_001284300.1 | NP_001271229.1 | 1 - 2 | 92 | 59 | ||
| NM_178058.4 | NP_835159.1 | 1 - 2 | 92 | 59 | ||
| GenBank mRNA | AK017403.1 | - | 1 - 2 | 90 | 59 | |
| AK085656.1 | - | 1 - 2 | 44 | 59 | ||
| AK147741.1 | - | 1 - 2 | 69 | 59 | ||
| AK155240.1 | - | 1 - 2 | 32 | 59 | ||
| BB847470.1 | - | 1 - 2 | 93 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK085656.1, AK147741.1, AK155240.1, NM_001039194.3, NM_001284300.1, NM_178058.4 transcripts.
Set Membership: |
|

Aifm2
Genomic Map