Assay Details
| Gene Symbol: Anxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007585.3 | NP_031611.1 | 8 - 9 | 665 | 73 | |
| GenBank mRNA | AK012563.1 | - | 9 - 10 | 667 | 73 | |
| AK088645.1 | - | 8 - 9 | 654 | 73 | ||
| AK146096.1 | - | 8 - 9 | 653 | 73 | ||
| AK150586.1 | - | 8 - 9 | 653 | 73 | ||
| AK152561.1 | - | 8 - 9 | 653 | 73 | ||
| AK160746.1 | - | 8 - 9 | 653 | 73 | ||
| AK164507.1 | - | 8 - 9 | 658 | 73 | ||
| AK165888.1 | - | 8 - 9 | 653 | 73 | ||
| AK167894.1 | - | 8 - 9 | 652 | 73 | ||
| AK168083.1 | - | 8 - 9 | 653 | 73 | ||
| BC003327.1 | - | 8 - 9 | 665 | 3156634 | 73 | |
| BC004659.1 | - | 4 - 5 | 1561 | 3497991 | 73 | |
| BC005763.1 | - | 8 - 9 | 639 | 2655513 | 73 | |
| D10024.1 | - | 7 - 8 | 598 | 73 | ||
| M14044.1 | - | 7 - 8 | 598 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa2
Genomic Map