Assay Details
| Gene Symbol: Trp63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127259.1 | NP_001120731.1 | 2 - 3 | 710 | 65 | |
| NM_001127260.1 | NP_001120732.1 | 2 - 3 | 710 | 65 | ||
| NM_001127261.1 | NP_001120733.1 | 2 - 3 | 710 | 65 | ||
| GenBank mRNA | AF075434.1 | - | 2 - 3 | 185 | 65 | |
| AF075435.1 | - | 2 - 3 | 185 | 65 | ||
| AF075436.1 | - | 2 - 3 | 185 | 65 | ||
| AK138305.1 | - | 2 - 3 | 714 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp63
Genomic Map