Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900007430.mm.1 |
NM_008613.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007430.mm.1 |
NM_008613.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007430.mm.1 |
TC0900000966.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007432.mm.1 |
NM_008613.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007432.mm.1 |
NM_008613.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007432.mm.1 |
TC0900000966.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007431.mm.1 |
NM_008613.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007431.mm.1 |
NM_008613.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007431.mm.1 |
TC0900000966.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007429.mm.1 |
NM_008613.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007429.mm.1 |
NM_008613.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007429.mm.1 |
TC0900000966.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007433.mm.1 |
NM_008613.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007433.mm.1 |
NM_008613.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007433.mm.1 |
TC0900000966.mm.1 |
MTA TC |