Assay Details
| Gene Symbol: Ap1m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110300.1 | NP_001103770.1 | 10 - 11 | 1333 | 64 | |
| NM_009678.2 | NP_033808.2 | 10 - 11 | 1327 | 64 | ||
| GenBank mRNA | AF067146.1 | - | 10 - 11 | 1301 | 64 | |
| AK010478.1 | - | 10 - 11 | 1312 | 64 | ||
| AK082674.1 | - | 1190 | 64 | |||
| BC003704.1 | - | 10 - 11 | 1266 | 3488231 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346492 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346494 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346495 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | BC003704.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | AK082674.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | AF067146.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | AK010478.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | NM_001110300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | BC003704.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | AK082674.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | AF067146.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | AK010478.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P638605 | NM_001110300.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970350 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35490 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151915 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149181 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | BC003704.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | AK082674.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | AF067146.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | AK010478.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | NM_001110300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | BC003704.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | AK082674.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | AF067146.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | AK010478.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P638605 | NM_001110300.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26346481 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06718325 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149181 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01881881 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06718336 | - | - |
| Whole Mouse Genome G4122A | A_52_P161845 | AK082674.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | BC003704.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | AK082674.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | AF067146.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | AK010478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | NM_001110300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | BC003704.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | AK082674.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | AF067146.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | AK010478.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P638605 | NM_001110300.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115738 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102612 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01881881 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149181 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08179668 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07631418 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07510281 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346495 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346494 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26346492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26346471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08037655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08188208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07540838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07925034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07540841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07611125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07631418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07920708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07920707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07631419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26346470 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591533 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591534 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591528 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591527 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460190_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591522 | - | - |
| Mouse Genome 430 2.0 Array | 1460190_at | - | - |
| Mouse Genome 430 2.0 Array | 1460190_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11768_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11768_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900015721.mm.1 | NM_001110300.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015721.mm.1 | NM_001110300.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015721.mm.1 | NM_009678.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015721.mm.1 | NM_009678.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015721.mm.1 | TC0900001868.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460190_PM_at | - | - |
| Mouse Expression Set 430 | 1460190_at | - | - |
| Murine Genome U74v2 Set | 135578_at | - | - |
| Murine Genome U74v2 Set | 135578_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460190_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460190_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460190_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460190_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6040324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2900138 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370537 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040324 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370537 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2762318 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2749011 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2619877 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2619877 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2762318 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2749011 | - | - |
| mouse-6 v1.1 (Illumina) | scl011768.1_215-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0011768.1_138-S | - | - |
| mouse-6 v1.1 (Illumina) | scl36145.7.8_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900138 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040324 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370537 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36145.7.8_1-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0011768.1_138-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl011768.1_215-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900138 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040324 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370537 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2925872 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2749011 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2619877 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2762318 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2619877 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2925872 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2749011 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2762318 | - | - |

Ap1m2
Genomic Map