Assay Details
| Gene Symbol: Ap1m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110300.1 | NP_001103770.1 | 3 - 4 | 357 | 74 | |
| NM_009678.2 | NP_033808.2 | 3 - 4 | 357 | 74 | ||
| GenBank mRNA | AK010478.1 | - | 3 - 4 | 336 | 74 | |
| AK082674.1 | - | 341 | 74 | |||
| AK160161.1 | - | 3 - 4 | 326 | 74 | ||
| BC003704.1 | - | 3 - 4 | 296 | 3488231 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap1m2
Genomic Map