Assay Details
| Gene Symbol: Ap1m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110300.1 | NP_001103770.1 | 4 - 5 | 633 | 99 | |
| NM_009678.2 | NP_033808.2 | 4 - 5 | 633 | 99 | ||
| GenBank mRNA | AF067146.1 | - | 4 - 5 | 607 | 99 | |
| AK010478.1 | - | 4 - 5 | 612 | 99 | ||
| BC003704.1 | - | 4 - 5 | 572 | 3488231 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap1m2
Genomic Map