| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900001309.mm.1 |
NM_001199186.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001309.mm.1 |
NM_001199186.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001309.mm.1 |
NM_152808.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001309.mm.1 |
NM_152808.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001309.mm.1 |
TC0900000193.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900001312.mm.1 |
NM_001199186.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001312.mm.1 |
NM_001199186.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001312.mm.1 |
NM_152808.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001312.mm.1 |
NM_152808.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900001312.mm.1 |
TC0900000193.mm.1 |
MTA TC |